accessibility menu, dialog, popup

UserName

H3CCH3OHO

CAS RN 88-09-5

IUPAC Name: 2-ethylbutanoic acid
Synonyms: 2-ethylbutyric acid diethyl acetic acid
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OXQGTIUCKGYOAA-UHFFFAOYSA-N
SMILES: CCC(CC)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)