missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylanisole 98.0+%, TCI America™
$137.94 - $415.68
Chemical Identifiers
| CAS | 14804-32-1 |
|---|---|
| Molecular Formula | C9H12O |
| Molecular Weight (g/mol) | 136.194 |
| MDL Number | MFCD00142909 |
| InChI Key | NIEHEMAZEULEKB-UHFFFAOYSA-N |
| Synonym | 2-Ethylmethoxybenzene |
| PubChem CID | 84654 |
| IUPAC Name | 1-ethyl-2-methoxybenzene |
| SMILES | CCC1=CC=CC=C1OC |
Chemical Identifiers
| 14804-32-1 | |
| 136.194 | |
| NIEHEMAZEULEKB-UHFFFAOYSA-N | |
| 84654 | |
| CCC1=CC=CC=C1OC |
| C9H12O | |
| MFCD00142909 | |
| 2-Ethylmethoxybenzene | |
| 1-ethyl-2-methoxybenzene |
Specifications
| 14804-32-1 | |
| 187°C | |
| MFCD00142909 | |
| 2-Ethylmethoxybenzene | |
| CCC1=CC=CC=C1OC | |
| 136.194 | |
| 136.19 | |
| Liquid |
| Yellow | |
| C9H12O | |
| 5 g | |
| NIEHEMAZEULEKB-UHFFFAOYSA-N | |
| 1-ethyl-2-methoxybenzene | |
| 84654 | |
| ≥98.0% (GC) | |
| 2-Ethylanisole |
Safety and Handling
EINECSNumber : (3)-0578&(3)-1035
TSCA : No