accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 14804-32-1

IUPAC Name: 1-ethyl-2-methoxybenzene
Synonyms: 2-ethylanisole
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: NIEHEMAZEULEKB-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results