missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanisole 98.0+%, TCI America™

Catalog No. E05095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05095G 5 g
E050925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05095G Supplier TCI America Supplier No. E05095G
Only null left

Chemical Identifiers

CAS 14804-32-1
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00142909
InChI Key NIEHEMAZEULEKB-UHFFFAOYSA-N
Synonym 2-Ethylmethoxybenzene
PubChem CID 84654
IUPAC Name 1-ethyl-2-methoxybenzene
SMILES CCC1=CC=CC=C1OC

Specifications

Color Yellow
Boiling Point 187°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylanisole
EINECSNumber (3)-0578&(3)-1035
TSCA No