missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%

CAS: 110-73-6 | C4H11NO | 89.138 g/mol

$71.66 - $247.14

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA15744AP
View Documents
Thermo Scientific Chemicals
A15744AP
500 mL
Each for $71.66
Only null left
 
AAA157440F
View Documents
Thermo Scientific Chemicals
A157440F
2500 mL
Each for $247.14
Only null left
 
Description

Description

2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

Solubility
Miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
Specifications

Chemical Identifiers

110-73-6
89.138
MIJDSYMOBYNHOT-UHFFFAOYSA-N
8072
CCNCCO
C4H11NO
MFCD00002841
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
2-(ethylamino)ethanol

Specifications

110-73-6
0.914
71°C (160°F)
1.441
MFCD00002841
UN2922
2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
MIJDSYMOBYNHOT-UHFFFAOYSA-N
2-(ethylamino)ethanol
8072
98%
-9°C
168°C to 170°C
C4H11NO
CH3CH2NHCH2CH2OH
500 mL
635671
Miscible in water.
CCNCCO
89.138
89.14
2-(Ethylamino)ethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H314-H318-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.

P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H311-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 203-797-5

RTECSNumber : KK9100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only