accessibility menu, dialog, popup

UserName

H3CNHOH

CAS RN 110-73-6

IUPAC Name: 2-(ethylamino)ethan-1-ol
Synonyms: ethyl ethanolamine
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: MIJDSYMOBYNHOT-UHFFFAOYSA-N
SMILES: CCNCCO
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)