Learn More
2-(Ethylamino)ethanol, 98%
CAS: 110-73-6 | C4H11NO | 89.138 g/mol
Supplier: Thermo Scientific Chemicals A157440F
Description
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.
Solubility
Miscible in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids.
Specifications
| 2-(Ethylamino)ethanol | |
| -9°C | |
| 168°C to 170°C | |
| C4H11NO | |
| CH3CH2NHCH2CH2OH | |
| 2500 mL | |
| 635671 | |
| Miscible in water. | |
| CCNCCO | |
| 89.138 | |
| 89.14 |
| 110-73-6 | |
| 0.914 | |
| 71°C (160°F) | |
| 1.441 | |
| MFCD00002841 | |
| UN2922 | |
| 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol | |
| MIJDSYMOBYNHOT-UHFFFAOYSA-N | |
| 2-(ethylamino)ethanol | |
| 8072 | |
| 98% |
Chemical Identifiers
| 110-73-6 | |
| 89.138 | |
| MIJDSYMOBYNHOT-UHFFFAOYSA-N | |
| 8072 | |
| CCNCCO |
| C4H11NO | |
| MFCD00002841 | |
| 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol | |
| 2-(ethylamino)ethanol |
Safety and Handling
GHS H Statement
H311-H314-H318-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H311-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 203-797-5
RTECSNumber : KK9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only