missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4,5-dimethylthiazole 98.0+%, TCI America™

$48.13 - $140.54

Chemical Identifiers

CAS 873-64-3
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00053125
InChI Key BVOKJOZBJOWZNJ-UHFFFAOYSA-N
PubChem CID 522877
IUPAC Name 2-ethyl-4,5-dimethyl-1,3-thiazole
SMILES CCC1=NC(=C(S1)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E07185G
View Documents
TCI America
E07185G
5 g
Each for $48.13
Only null left
 
E071825G
View Documents
TCI America
E071825G
25 g
Each for $140.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

873-64-3
141.232
BVOKJOZBJOWZNJ-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-thiazole
C7H11NS
MFCD00053125
522877
CCC1=NC(=C(S1)C)C

Specifications

873-64-3
185°C
MFCD00053125
BVOKJOZBJOWZNJ-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-thiazole
522877
≥98.0% (GC)
2-Ethyl-4,5-dimethylthiazole
Yellow
C7H11NS
5 g
CCC1=NC(=C(S1)C)C
141.232
141.23
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents