missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-4,5-dimethylthiazole 98.0+%, TCI America™
Supplier: TCI America E071825G
Specifications
| 2-Ethyl-4,5-dimethylthiazole | |
| Yellow | |
| C7H11NS | |
| 25 g | |
| CCC1=NC(=C(S1)C)C | |
| 141.232 | |
| 141.23 | |
| Liquid |
| 873-64-3 | |
| 185°C | |
| MFCD00053125 | |
| BVOKJOZBJOWZNJ-UHFFFAOYSA-N | |
| 2-ethyl-4,5-dimethyl-1,3-thiazole | |
| 522877 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 873-64-3 | |
| 141.232 | |
| BVOKJOZBJOWZNJ-UHFFFAOYSA-N | |
| 2-ethyl-4,5-dimethyl-1,3-thiazole |
| C7H11NS | |
| MFCD00053125 | |
| 522877 | |
| CCC1=NC(=C(S1)C)C |
Safety and Handling
TSCA : No