missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-2-methyl-1,3-propanediol 97.0+%, TCI America™
Supplier: TCI America E041225G
Specifications
| 2-Ethyl-2-methyl-1,3-propanediol | |
| 43°C | |
| 226°C | |
| MFCD00004693 | |
| 2,2-Bis(hydroxymethyl)butane | |
| CCC(C)(CO)CO | |
| 118.176 | |
| 118.18 | |
| Crystalline Lumps |
| 77-84-9 | |
| White-Yellow | |
| C6H14O2 | |
| 25 g | |
| VNAWKNVDKFZFSU-UHFFFAOYSA-N | |
| 2-ethyl-2-methylpropane-1,3-diol | |
| 66169 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 77-84-9 | |
| 118.176 | |
| VNAWKNVDKFZFSU-UHFFFAOYSA-N | |
| 66169 | |
| CCC(C)(CO)CO |
| C6H14O2 | |
| MFCD00004693 | |
| 2,2-Bis(hydroxymethyl)butane | |
| 2-ethyl-2-methylpropane-1,3-diol |
Safety and Handling
TSCA : No