missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-methyl-1,3-propanediol 97.0+%, TCI America™

Catalog No. p-7128986
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04125G 5 g
E041225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04125G Supplier TCI America Supplier No. E04125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-84-9
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004693
InChI Key VNAWKNVDKFZFSU-UHFFFAOYSA-N
Synonym 2,2-Bis(hydroxymethyl)butane
PubChem CID 66169
IUPAC Name 2-ethyl-2-methylpropane-1,3-diol
SMILES CCC(C)(CO)CO

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 226°C
Quantity 5 g
Formula Weight 118.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Ethyl-2-methyl-1,3-propanediol