accessibility menu, dialog, popup

UserName

CH3CH3HOHO

CAS RN 77-84-9

IUPAC Name: 2-ethyl-2-methylpropane-1,3-diol
Synonyms: 2-ethyl-2-methylpropane-1,3-diol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: VNAWKNVDKFZFSU-UHFFFAOYSA-N
SMILES: CCC(C)(CO)CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results