missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-1-hexanol 99.5+%, TCI America™
$36.07 - $49.36
Chemical Identifiers
| CAS | 104-76-7 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.231 |
| MDL Number | MFCD00004746 |
| InChI Key | YIWUKEYIRIRTPP-UHFFFAOYSA-N |
| Synonym | 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 |
| PubChem CID | 7720 |
| ChEBI | CHEBI:16011 |
| IUPAC Name | 2-ethylhexan-1-ol |
| SMILES | CCCCC(CC)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
E012225ML
|
N/A | 25 mL |
Each for $36.07
|
|
|||||
|
E0122500ML
|
N/A | 500 mL |
Each for $49.36
|
|
|||||
Chemical Identifiers
| C8H18O | |
| MFCD00004746 | |
| 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 | |
| CHEBI:16011 | |
| CCCCC(CC)CO |
Specifications
| 104-76-7 | |
| 187°C | |
| MFCD00004746 | |
| YIWUKEYIRIRTPP-UHFFFAOYSA-N | |
| 2-ethylhexan-1-ol | |
| 7720 | |
| 130.23 | |
| Liquid |
| Colorless | |
| C8H18O | |
| 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 | |
| CCCCC(CC)CO | |
| 130.231 | |
| CHEBI:16011 | |
| ≥99.5% (GC) | |
| 2-Ethyl-1-hexanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MP0350000
TSCA : Yes