missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-hexanol 99.5+%, TCI America™

Catalog No. E012225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E012225ML 25 mL
E0122500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E012225ML Supplier TCI America Supplier No. E012225ML

Chemical Identifiers

CAS 104-76-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004746
InChI Key YIWUKEYIRIRTPP-UHFFFAOYSA-N
Synonym 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151
PubChem CID 7720
ChEBI CHEBI:16011
IUPAC Name 2-ethylhexan-1-ol
SMILES CCCCC(CC)CO

Specifications

Color Colorless
Boiling Point 187°C
Quantity 25 mL
Formula Weight 130.23
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-hexanol
EINECSNumber (2)-0217
RTECSNumber MP0350000
TSCA Yes