missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Acetate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor E102420LTMT

 View more versions of this product

Catalog No. 18608212


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

0.0002
1
20 L
Reagent
111-15-9
Steel Can
0.99

Chemical Identifiers

111-15-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.