missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethyl Acetate, 99%, Spectrum™ Chemical
Specifications
Specifications
| Acidity | 0.0002 |
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 1 L |
| Percent Purity | 0.99 |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.