accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 111-15-9

IUPAC Name: 2-ethoxyethyl acetate
Synonyms: 2-ethoxyethyl acetate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: SVONRAPFKPVNKG-UHFFFAOYSA-N
SMILES: CCOCCOC(C)=O
Boiling Point 
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)