missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether), Reagents
2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C21992004C
Specifications
| 2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether) | |
| 135°C | |
| Colorless | |
| -70°C | |
| C4H10O2 | |
| 0.93 | |
| ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
| 2-ethoxyethan-1-ol | |
| Reagent |
| 110-80-5 | |
| -70°C | |
| 135°C | |
| Glass Bottle | |
| 4 L | |
| Ethylene Glycol Monoethyl Ether, Cellosolve<superscriptT><superscriptM> | |
| CCOCCO | |
| 90.12 | |
| Liquid |
Chemical Identifiers
| 110-80-5 | |
| 90.12 | |
| Ethylene Glycol Monoethyl Ether, Cellosolve<superscriptT><superscriptM> | |
| CCOCCO |
| C4H10O2 | |
| ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
| 2-ethoxyethan-1-ol |