missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether), Reagents

Catalog No. p-200021218
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337267 1 L
01337266 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337267 Supplier Labchem Inc Supplier No. C21992001C
Only null left

2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym Ethylene Glycol Monoethyl Ether, Cellosolve<superscriptT><superscriptM>
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Boiling Range 135°C
Melting Point -70°C
Color Colorless
Boiling Point 135°C
Melting Range -70°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.93
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethanol (Ethylene Glycol Monoethyl Ether)
Show More Show Less