Learn More
2-Ethoxycarbonylphenylboronic acid, 97%, Thermo Scientific™
$197.76 - $731.31
Chemical Identifiers
| CAS | 380430-53-5 |
|---|---|
| Molecular Formula | C9H11BO4 |
| Molecular Weight (g/mol) | 193.99 |
| MDL Number | MFCD02179453 |
| InChI Key | QZKVVOXAEBCLPZ-UHFFFAOYSA-N |
| Synonym | 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid |
| PubChem CID | 2773405 |
| IUPAC Name | (2-ethoxycarbonylphenyl)boronic acid |
| SMILES | CCOC(=O)C1=CC=CC=C1B(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC393830050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC393830250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 380430-53-5 | |
| 193.99 | |
| QZKVVOXAEBCLPZ-UHFFFAOYSA-N | |
| 2773405 | |
| CCOC(=O)C1=CC=CC=C1B(O)O |
| C9H11BO4 | |
| MFCD02179453 | |
| 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid | |
| (2-ethoxycarbonylphenyl)boronic acid |
Specifications
| 380430-53-5 | |
| Glass bottle | |
| MFCD02179453 | |
| Solubility in water: .. Other solubilities: soluble in methanol | |
| CCOC(=O)C1=CC=CC=C1B(O)O | |
| 193.99 | |
| 193.99 | |
| 2-Ethoxycarbonylphenylboronic acid |
| 96% min. (HPLC) | |
| C9H11BO4 | |
| 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid | |
| QZKVVOXAEBCLPZ-UHFFFAOYSA-N | |
| (2-ethoxycarbonylphenyl)boronic acid | |
| 2773405 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medi
GHS Signal Word: Warning
RUO – Research Use Only