missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxycarbonylphenylboronic acid, 97%

Catalog No. AC393830050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC393830050 5 g
AC393830250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393830050 Supplier Thermo Scientific Chemicals Supplier No. 393830050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 380430-53-5 | C9H11BO4 | 193.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-53-5
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179453
InChI Key QZKVVOXAEBCLPZ-UHFFFAOYSA-N
Synonym 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid
PubChem CID 2773405
IUPAC Name (2-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 137.0°C to 138.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 193.99
Percent Purity 97%
Chemical Name or Material 2-Ethoxycarbonylphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medi

RUO – Research Use Only