missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7129032
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
E04995G 5 g
E049925G 25 g
2 options

Catalog No. E04995G

Supplier: TCI America E04995G

Only null left

Chemical Identifiers

CAS 71672-75-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004616
InChI Key ICJVQAHPHKYCNU-UHFFFAOYSA-N
Synonym 2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn
PubChem CID 51381
IUPAC Name (2-ethoxyphenyl)methanol
SMILES CCOC1=CC=CC=C1CO

Specifications

Color Yellow
Boiling Point 265°C
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzyl Alcohol
RTECSNumber DO5467000
TSCA No