accessibility menu, dialog, popup

UserName

CH3OOH

CAS RN 71672-75-8

IUPAC Name: (2-ethoxyphenyl)methanol
Synonyms: (2-ethoxyphenyl)methanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: ICJVQAHPHKYCNU-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1CO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)