missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzyl alcohol, 98%

Catalog No. AAA1547722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547722 100 g
AAA1547714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1547722 Supplier Thermo Scientific Chemicals Supplier No. A1547722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71672-75-8 | C9H12O2 | 152.19 g/mol

2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product. It is also used as pharmaceutical intermediates.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 71672-75-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004616
InChI Key ICJVQAHPHKYCNU-UHFFFAOYSA-N
Synonym 2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn
PubChem CID 51381
IUPAC Name (2-ethoxyphenyl)methanol
SMILES CCOC1=CC=CC=C1CO

Specifications

Density 1.074
Boiling Point 263°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 100 g
Beilstein 2555374
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-Ethoxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 275-811-8
RTECSNumber DO5467000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only