missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxy-5-(1-propenyl)phenol 98.0+%, TCI America™
$73.16 - $272.03
Chemical Identifiers
| CAS | 94-86-0 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.23 |
| MDL Number | MFCD00009283 |
| InChI Key | RADIRXJQODWKGQ-HWKANZROSA-N |
| Synonym | vanitrope, propenylguaethol, 6-ethoxy-m-anol, isosafroeugenol, 2-ethoxy-5-1-propenyl phenol, hydroxy methyl anethol, propenyl guaethol, 2-ethoxy-5-propenylphenol, trans-2-ethoxy-5-1-propenyl phenol, phenol, 2-ethoxy-5-propenyl |
| PubChem CID | 5354280 |
| IUPAC Name | 2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol |
| SMILES | CCOC1=CC=C(\C=C\C)C=C1O |
Chemical Identifiers
| 94-86-0 | |
| 178.23 | |
| RADIRXJQODWKGQ-HWKANZROSA-N | |
| 5354280 | |
| CCOC1=CC=C(\C=C\C)C=C1O |
| C11H14O2 | |
| MFCD00009283 | |
| vanitrope, propenylguaethol, 6-ethoxy-m-anol, isosafroeugenol, 2-ethoxy-5-1-propenyl phenol, hydroxy methyl anethol, propenyl guaethol, 2-ethoxy-5-propenylphenol, trans-2-ethoxy-5-1-propenyl phenol, phenol, 2-ethoxy-5-propenyl | |
| 2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol |
Specifications
| 94-86-0 | |
| White | |
| C11H14O2 | |
| 25 g | |
| RADIRXJQODWKGQ-HWKANZROSA-N | |
| 2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol | |
| 5354280 | |
| ≥98.0% (GC) | |
| 2-Ethoxy-5-(1-propenyl)phenol |
| 86°C | |
| 116°C | |
| MFCD00009283 | |
| vanitrope, propenylguaethol, 6-ethoxy-m-anol, isosafroeugenol, 2-ethoxy-5-1-propenyl phenol, hydroxy methyl anethol, propenyl guaethol, 2-ethoxy-5-propenylphenol, trans-2-ethoxy-5-1-propenyl phenol, phenol, 2-ethoxy-5-propenyl | |
| CCOC1=CC=C(\C=C\C)C=C1O | |
| 178.23 | |
| 178.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0636
RTECSNumber : SL3850000
TSCA : Yes