missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-5-(1-propenyl)phenol 98.0+%, TCI America™

Supplier:  TCI America E0804100G

 View more versions of this product

Catalog No. E0804100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Ethoxy-5-(1-propenyl)phenol
86°C
116°C
MFCD00009283
vanitrope, propenylguaethol, 6-ethoxy-m-anol, isosafroeugenol, 2-ethoxy-5-1-propenyl phenol, hydroxy methyl anethol, propenyl guaethol, 2-ethoxy-5-propenylphenol, trans-2-ethoxy-5-1-propenyl phenol, phenol, 2-ethoxy-5-propenyl
CCOC1=CC=C(\C=C\C)C=C1O
178.23
178.23
Crystalline Powder
94-86-0
White
C11H14O2
100 g
RADIRXJQODWKGQ-HWKANZROSA-N
2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol
5354280
≥98.0% (GC)

Chemical Identifiers

94-86-0
178.23
RADIRXJQODWKGQ-HWKANZROSA-N
5354280
CCOC1=CC=C(\C=C\C)C=C1O
C11H14O2
MFCD00009283
vanitrope, propenylguaethol, 6-ethoxy-m-anol, isosafroeugenol, 2-ethoxy-5-1-propenyl phenol, hydroxy methyl anethol, propenyl guaethol, 2-ethoxy-5-propenylphenol, trans-2-ethoxy-5-1-propenyl phenol, phenol, 2-ethoxy-5-propenyl
2-ethoxy-5-[(1E)-prop-1-en-1-yl]phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0636

RTECSNumber : SL3850000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions