missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecylphenol 98.0+%, TCI America™

Catalog No. D230210G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D230210G 10 g
D230225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D230210G Supplier TCI America Supplier No. D230210G

Chemical Identifiers

CAS 5284-29-7
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.44
MDL Number MFCD00039521,MFCD00020106
InChI Key CYEJMVLDXAUOPN-UHFFFAOYSA-N
Synonym 2-Laurylphenol
PubChem CID 171144
IUPAC Name 2-dodecylphenol
SMILES CCCCCCCCCCCCC1=CC=CC=C1O

Specifications

Melting Point 46°C
Color White
Quantity 10 g
Formula Weight 262.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Dodecylphenol