accessibility menu, dialog, popup

UserName

H3CHO

CAS RN 5284-29-7

IUPAC Name: 2-dodecylphenol
Synonyms: 2-dodecylphenol
Molecular Weight (g/mol): 262.44
Molecular Formula: C18H30O
InChi Key: CYEJMVLDXAUOPN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results