missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzoic Acid 98.0+%, TCI America™

Catalog No. p-7127637
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32421G 1 g
D32425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32421G Supplier TCI America Supplier No. D32421G

Chemical Identifiers

CAS 17261-28-8
Molecular Formula C19H15O2P
Molecular Weight (g/mol) 306.30
MDL Number MFCD00674024
InChI Key UYRPRYSDOVYCOU-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzoic acid, 2-diphenylphosphinobenzoic acid, benzoic acid, 2-diphenylphosphino, o-diphenylphosphinobenozic acid, 2-carboxyphenyl diphenylphosphine, 2-diphenylphosphanyl benzoic acid, benzoic acid, diphenylphosphino, o-diphenylphosphino benzoic acid, dppbac, acmc-1bt7u
PubChem CID 87021
IUPAC Name 2-(diphenylphosphanyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 180°C
Color Yellow
Quantity 1 g
Formula Weight 306.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diphenylphosphino)benzoic Acid
RTECSNumber DG9287500
TSCA Yes