missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzaldehyde 97.0+%, TCI America™

Catalog No. p-7128260
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44111G 1 g
D44115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44111G Supplier TCI America Supplier No. D44111G
Only null left

Chemical Identifiers

CAS 50777-76-9
Molecular Formula C19H15OP
Molecular Weight (g/mol) 290.30
MDL Number MFCD00013367
InChI Key DRCPJRZHAJMWOU-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzaldehyde, 2-diphenylphosphinobenzaldehyde, o-diphenylphosphino benzaldehyde, 2-formylphenyl diphenylphosphine, 2-diphenylphosphanyl benzaldehyde, benzaldehyde, 2-diphenylphosphino, dppbde, zlchem 984, 2-formylphenyldiphenylphosphine
PubChem CID 2754316
IUPAC Name 2-(diphenylphosphanyl)benzaldehyde
SMILES O=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 117°C
Color Yellow
Quantity 1 g
Formula Weight 290.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde