accessibility menu, dialog, popup

UserName

CAS RN 50777-76-9

IUPAC Name: 2-(diphenylphosphanyl)benzaldehyde
Synonyms: 2-(diphenylphosphanyl)benzaldehyde
Molecular Weight (g/mol): 290.3
Molecular Formula: C19H15OP
InChi Key: DRCPJRZHAJMWOU-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)