missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Acetate 98.0+%, TCI America™

Supplier:  TCI America A0929500ML

 View more versions of this product

Catalog No. A0929500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(Dimethylamino)ethyl Acetate
Yellow
C6H13NO2
500 mL
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
CN(C)CCOC(C)=O
131.18
131.18
Liquid
1421-89-2
152°C
MFCD00041912
1993
GOLSFPMYASLXJC-UHFFFAOYSA-N
2-(dimethylamino)ethyl acetate
15013
≥98.0% (GC,T)

Chemical Identifiers

1421-89-2
131.18
GOLSFPMYASLXJC-UHFFFAOYSA-N
15013
CN(C)CCOC(C)=O
C6H13NO2
MFCD00041912
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
2-(dimethylamino)ethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AH2100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions