missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Acetate 98.0+%, TCI America™

Catalog No. p-7117135
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A092925ML 25 mL
A0929500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A092925ML Supplier TCI America Supplier No. A092925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1421-89-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00041912
InChI Key GOLSFPMYASLXJC-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate
PubChem CID 15013
IUPAC Name 2-(dimethylamino)ethyl acetate
SMILES CN(C)CCOC(C)=O

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 mL
UN Number 1993
Formula Weight 131.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl Acetate
RTECSNumber AH2100000
TSCA Yes