missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxyinosine 98.0+%, TCI America™
Supplier: TCI America D35845G
Specifications
| 2′-Deoxyinosine | |
| 220°C | |
| C10H12N4O4 | |
| 5 g | |
| VGONTNSXDCQUGY-YUZWJPFSNA-N | |
| 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one | |
| 65058 | |
| 252.23 | |
| Crystalline Powder |
| 890-38-0 | |
| White | |
| MFCD00005762 | |
| 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O | |
| 252.23 | |
| CHEBI:28997 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 890-38-0 | |
| 252.23 | |
| VGONTNSXDCQUGY-YUZWJPFSNA-N | |
| 65058 | |
| 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| C10H12N4O4 | |
| MFCD00005762 | |
| 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine | |
| CHEBI:28997 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O |
Safety and Handling
TSCA : No