missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyinosine 98.0+%, TCI America™

Catalog No. p-7127803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35841G 1 g
D35845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35841G Supplier TCI America Supplier No. D35841G

Chemical Identifiers

CAS 890-38-0
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00005762
InChI Key VGONTNSXDCQUGY-YUZWJPFSNA-N
Synonym 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
PubChem CID 65058
ChEBI CHEBI:28997
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 220°C
Color White
Quantity 1 g
Formula Weight 252.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyinosine