missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine-5'-monophosphate, 99%

CAS: 1032-65-1 | C9H14N3O7P | 307.199 g/mol

Supplier:  Thermo Scientific Chemicals J6524406

 View more versions of this product

Catalog No. AAJ6524406


Only null left
Explore available promotions

Description

Description

2'-Deoxycytidine-5'-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine-5′-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2'-Deoxycytidine-5'-monophosphate
White
MFCD00006546
41062
Soluble in water.
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O
307.199
CHEBI:15918
99%
1032-65-1
C9H14N3O7P
5 g
dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
NCMVOABPESMRCP-SHYZEUOFSA-N
[(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
13945
307.2
Crystalline Powder

Chemical Identifiers

1032-65-1
307.199
NCMVOABPESMRCP-SHYZEUOFSA-N
13945
[(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
C9H14N3O7P
MFCD00006546
dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
CHEBI:15918
C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-849-9

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only