missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine-5'-monophosphate, 99%

Catalog No. p-7043049
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6524403 1 g
AAJ6524406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6524403 Supplier Thermo Scientific Chemicals Supplier No. J6524403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1032-65-1 | C9H14N3O7P | 307.199 g/mol

2'-Deoxycytidine-5'-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxycytidine-5′-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1032-65-1
Molecular Formula C9H14N3O7P
Molecular Weight (g/mol) 307.199
MDL Number MFCD00006546
InChI Key NCMVOABPESMRCP-SHYZEUOFSA-N
Synonym dcmp, deoxycytidylic acid, deoxycytidine monophosphate, 2'-deoxycytidine-5'-monophosphate, 2'-deoxycytidine-5'-monophosphoric acid, 5'-cytidylic acid, 2'-deoxy, 2'-deoxycytidine 5'-monophosphate, deoxycytidylate, polydeoxycytidylic acid, deoxycytidine-5'-monophosphoric acid
PubChem CID 13945
ChEBI CHEBI:15918
IUPAC Name [(2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1C(C(OC1N2C=CC(=NC2=O)N)COP(=O)(O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 41062
Solubility Information Soluble in water.
Formula Weight 307.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine-5'-monophosphate
EINECSNumber 213-849-9
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only