accessibility menu, dialog, popup

UserName

CAS RN 1032-65-1

IUPAC Name: {[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: deoxycytidylate 2'-deoxycytidine-5'-phosphate deoxycytidine-phosphate monophosphate, deoxycytidine deoxycytidylic acids acid, deoxycytidylic deoxycytidylic acid (substance) deoxycytidylic acid acids, deoxycytidylic DCMP
Molecular Weight (g/mol): 307.2
Molecular Formula: C9H14N3O7P
InChi Key: NCMVOABPESMRCP-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1)C1CC(O)C(COP(O)(O)=O)O1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)