missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-ribose 98.0+%, TCI America™

Supplier:  TCI America D00595G

 View more versions of this product

Catalog No. D00595G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Deoxy-D-ribose
90°C
C5H10O4
5 g
PDWIQYODPROSQH-GLULIZFNNA-N
(4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
5460005
134.13
Crystalline Powder
533-67-5
White-Yellow
MFCD00135904
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
OC[C@H]1OC(O)C[C@@H]1O
134.13
CHEBI:28816
≥98.0% (HPLC)

Chemical Identifiers

533-67-5
134.13
PDWIQYODPROSQH-GLULIZFNNA-N
5460005
(4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
C5H10O4
MFCD00135904
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
CHEBI:28816
OC[C@H]1OC(O)C[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6008

RTECSNumber : SB7230000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions