missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-ribose 98.0+%, TCI America™

Catalog No. p-7126005
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00595G 5 g
D005925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00595G Supplier TCI America Supplier No. D00595G
Only null left

Chemical Identifiers

CAS 533-67-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00135904
InChI Key PDWIQYODPROSQH-GLULIZFNNA-N
Synonym 2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PubChem CID 5460005
ChEBI CHEBI:28816
IUPAC Name (4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
SMILES OC[C@H]1OC(O)C[C@@H]1O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 134.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-D-ribose
EINECSNumber (5)-6008
RTECSNumber SB7230000
TSCA Yes