accessibility menu, dialog, popup

UserName

CAS RN 533-67-5

(S)(R)OHOHOOH

CAS RN 533-67-5

IUPAC Name: (4S,5R)-5-(hydroxymethyl)oxolane-2,4-diol
Synonyms: 2-deoxy-D-ribofuranose
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: PDWIQYODPROSQH-GLULIZFNNA-N
SMILES: OC[C@H]1OC(O)C[C@@H]1O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (4)