missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxy-5-(hydroxymethyl)cytidine 98.0+%, TCI America™
Supplier: TCI America D422050MG
Specifications
| 2′-Deoxy-5-(hydroxymethyl)cytidine | |
| 205°C | |
| MFCD09842113 | |
| 5-hmdC | |
| C1C(C(OC1N2C=C(C(=NC2=O)N)CO)CO)O | |
| 257.246 | |
| 257.25 | |
| Crystalline Powder |
| 7226-77-9 | |
| C10H15N3O5 | |
| 50 mg | |
| HMUOMFLFUUHUPE-CSMHCCOUSA-N | |
| 4-amino-1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one | |
| 98007919 | |
| ≥98.0% (T) |
Chemical Identifiers
| 7226-77-9 | |
| 257.246 | |
| HMUOMFLFUUHUPE-CSMHCCOUSA-N | |
| 98007919 | |
| C1C(C(OC1N2C=C(C(=NC2=O)N)CO)CO)O |
| C10H15N3O5 | |
| MFCD09842113 | |
| 5-hmdC | |
| 4-amino-1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one |
Safety and Handling
TSCA : No