accessibility menu, dialog, popup

UserName

CAS RN 7226-77-9

IUPAC Name: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one
Molecular Weight (g/mol): 257.25
Molecular Formula: C10H15N3O5
InChi Key: HMUOMFLFUUHUPE-XLPZGREQSA-N
SMILES: NC1=NC(=O)N(C=C1CO)[C@H]1C[C@H](O)[C@@H](CO)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results