missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-5-(hydroxymethyl)cytidine 98.0+%, TCI America™

Catalog No. p-7128161
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D422050MG 50 mg
D4220200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D422050MG Supplier TCI America Supplier No. D422050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7226-77-9
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.246
MDL Number MFCD09842113
InChI Key HMUOMFLFUUHUPE-CSMHCCOUSA-N
Synonym 5-hmdC
PubChem CID 98007919
IUPAC Name 4-amino-1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)CO)CO)O

Specifications

Melting Point 205°C
Quantity 50 mg
Formula Weight 257.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-5-(hydroxymethyl)cytidine