missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylphenol 97.0+%, TCI America™

Catalog No. p-7124581
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07325G 5 g
C073225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07325G Supplier TCI America Supplier No. C07325G
Only null left

Chemical Identifiers

CAS 119-42-6
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00019335
InChI Key MVRPPTGLVPEMPI-UHFFFAOYSA-N
Synonym 2-Hydroxyphenylcyclohexane
PubChem CID 8396
IUPAC Name 2-cyclohexylphenol
SMILES C1CCC(CC1)C2=CC=CC=C2O

Specifications

Melting Point 56°C
Color Red-White
Boiling Point 284°C
Quantity 5 g
Formula Weight 176.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyclohexylphenol
EINECSNumber (4)-0021
TSCA Yes