missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylbenzoic acid, 97%

Catalog No. p-7047217
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1806003 1 g
AAL1806006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1806003 Supplier Thermo Scientific Chemicals Supplier No. L1806003

CAS: 97023-48-8 | C13H16O2 | 204.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97023-48-8
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00452653
InChI Key ZKTFZNPTAJIXMK-UHFFFAOYSA-N
Synonym benzoic acid, 2-cyclohexyl, cyclohexylbenzoic acid, 2-cyclohexyl benzoic acid, 2-cyclohexyl-benzoic acid
PubChem CID 7020572
IUPAC Name 2-cyclohexylbenzoic acid
SMILES C1CCC(CC1)C2=CC=CC=C2C(=O)O

Specifications

Melting Point 103°C to 105°C
Quantity 1 g
Beilstein 2452158
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 2-Cyclohexylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only