missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyanothiophene 98.0+%, TCI America™
Supplier: TCI America T212125G
Specifications
| 2-Cyanothiophene | |
| Yellow | |
| C5H3NS | |
| 25 g | |
| 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile | |
| C1=CSC(=C1)C#N | |
| 109.146 | |
| 109.15 | |
| Liquid |
| 1003-31-2 | |
| 192°C | |
| MFCD00005416 | |
| 1992 | |
| CUPOOAWTRIURFT-UHFFFAOYSA-N | |
| thiophene-2-carbonitrile | |
| 66087 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1003-31-2 | |
| 109.146 | |
| CUPOOAWTRIURFT-UHFFFAOYSA-N | |
| 66087 | |
| C1=CSC(=C1)C#N |
| C5H3NS | |
| MFCD00005416 | |
| 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile | |
| thiophene-2-carbonitrile |
Safety and Handling
TSCA : No