missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothiophene 98.0+%, TCI America™

Catalog No. p-7137095
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21215G 5 g
T212125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21215G Supplier TCI America Supplier No. T21215G
Only null left

Chemical Identifiers

CAS 1003-31-2
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00005416
InChI Key CUPOOAWTRIURFT-UHFFFAOYSA-N
Synonym 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
PubChem CID 66087
IUPAC Name thiophene-2-carbonitrile
SMILES C1=CSC(=C1)C#N

Specifications

Color Yellow
Boiling Point 192°C
Quantity 5 g
UN Number 1992
Formula Weight 109.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyanothiophene