accessibility menu, dialog, popup

UserName

NS

CAS RN 1003-31-2

IUPAC Name: thiophene-2-carbonitrile
Synonyms: 2-cyanothiophene
Molecular Weight (g/mol): 109.15
Molecular Formula: C5H3NS
InChi Key: CUPOOAWTRIURFT-UHFFFAOYSA-N
SMILES: N#CC1=CC=CS1
Boiling Point 
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)