missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothioacetamide, 97%

Catalog No. AC305680050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305680050 5 g Glass bottle
AC305680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305680050 Supplier Thermo Scientific Chemicals Supplier No. 305680050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7357-70-2 | C3H4N2S | 100.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7357-70-2
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00010025
InChI Key BHPYMZQTCPRLNR-UHFFFAOYSA-N
Synonym 2-cyanothioacetamide, cyanothioacetamide, ethanethioamide, 2-cyano, cyano thioacetamide, 2-cyano-thioacetamide, 3-amino-3-thioxopropanenitrile, cyanthioacetamide, cyanoacetothioamide, 2-cyanothioactamide, alpha-cyanothioacetamide
PubChem CID 1416277
IUPAC Name 2-cyanoethanethioamide
SMILES NC(=S)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C to 119°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCCH2CSNH2
Quantity 5 g
Formula Weight 100.14
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 2-Cyanothioacetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only